Would you like to log in again?
Mini Q, GE Healthcare
Supplier: GE Healthcare - Life Sciences
Mini Q™
17-0686-01 17-0686-01 17-5177-01 17-5177-01
95016-950EA 2337.07 USD InStock
CA95016-950L 95016-950 CA95056-272L 95056-272
Mini Q, GE Healthcare
Chromatography Columns
Mini Q 4.6/50 PE columns are prepacked with MiniBeads and are designed for high-performance purification and characterization of biomolecules by ion exchange chromatography
  • Monodisperse 3 µm nonporous beads yield exceptional resolution, speed, reproducibility, and durability.
  • PC columns are designed for micropurification.
  • Nonporous matrix of MiniBeads improves resolution and flow kinetics, better withstands back pressure, and minimizes nonspecific adsorption of the sample.
  • Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q and S ion exchangers.
  • Nonporous matrix of MiniBeads improves resolution and flow kinetics, better withstands back pressure, and minimizes nonspecific adsorption of the sample.
Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q and S ion exchangers. Precision Column Holder enables use of precision columns with high-performance chromatography systems such as ÄKTA design.
The small particle size of MiniBeads offers the highest resolution. Precision Column Holder enables use of precision columns with high-performance chromatography systems such as ÄKTA design.
Delivery information: Includes Valco fittings for ÄKTAdesign systems and are supplied with M6 connectors for FPLC System.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty