Would you like to log in again?
Mono S GL Columns, GE Healthcare
Supplier: GE Healthcare - Life Sciences
Mono S™
17-5180-01 17-5180-01 17-5169-01 17-5169-01 17-5168-01 17-5168-01
95055-898EA 2383.65 USD InStock
95055-898 CA95055-898L 95056-270 CA95056-270L 95056-268 CA95056-268L
Mono S GL Columns, GE Healthcare
Chromatography Columns
Monodisperse 10 µm porous beads yield high resolution, dynamic capacity, reproducibility, and durability
  • MonoBeads are also available as Mono P for chromatofocusing
  • Compatible with ÄKTA design, FPLC System, and other high-performance LC systems
  • Not suitable for use with ÄKTAprime plus system
Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the S ion exchangers. Valco fittings for ÄKTAdesign systems and are supplied with M6 connectors for FPLC System.
Delivery information: Includes Valco fittings for ÄKTAdesign systems and are supplied with M6 connectors for FPLC System.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty