Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
HR Mono Q, GE Healthcare
Supplier: GE Healthcare - Life Sciences
17050601 17050601
97067-928EA 14754 USD InStock
CA97067-928 97067-928
HR Mono Q, GE Healthcare
Chromatography Columns
Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q ion exchangers
  • Monodisperse 10 µm porous beads yield high resolution, dynamic capacity, reproducibility, and durability
  • Compatible with ÄKTA design, FPLC System, and other high-performance LC systems
  • Not suitable for use with ÄKTAprime plus system
MonoBeads are also available as Mono P for chromatofocusing.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty