Sepharose™ Gel Filtration Base Matrix, Cytiva
Supplier: Cytiva
Sepharose™
17-0150-01 17-0120-01 17-0140-01 17-0110-01 17-0160-01 17-0160-01 17-0110-01 17-0150-01 17-0120-01 17-0140-01
95056-224EA 1171.18 USD
CA95056-224L CA95056-028L CA95056-138L CA95056-026L 95056-224 95056-026 95056-138 95056-028 CA95056-222L 95056-222
Sepharose™ Gel Filtration Base Matrix, Cytiva
Protein Purification Protein Purification Resins
Sepharose is a bead-formed agarose-based gel filtration matrix.
  • Proven base matrix for coupling affinity ligands
  • Higher degree of crosslinking improve matrix stability
Sepharose is a bead-formed agarose-based gel filtration matrix. Sepharose is available with 3 different agarose contents; 2, 4, and 6%, designated Sepharose 2B, Sepharose 4B and Sepharose 6B respectively, Both Sepharose and Sepharose CL have broad fractionation ranges which makes them suitable for characterizing or cleaning-up samples containing components of diverse molecular weight. Sepharose CL gels are cross-linked derivatives of Sepharose 2B, Sepharose 4B and Sepharose 6B. The cross-linked form of Sepharose is chemically and physically more resistant than Sepharose itself, offering the same selectivity with better flow characteristics. Cross-linked Sepharose gels are resistant to organic solvents and are thus the choice for separations in organic solvents.
Sepharose is available with 3 different agarose contents; 2, 4, and 6%, designated Sepharose 2B, Sepharose 4B and Sepharose 6B respectively, Both Sepharose and Sepharose CL have broad fractionation ranges which makes them suitable for characterizing or cleaning-up samples containing components of diverse molecular weight. Sepharose CL gels are cross-linked derivatives of Sepharose 2B, Sepharose 4B and Sepharose 6B. The cross-linked form of Sepharose is chemically and physically more resistant than Sepharose itself, offering the same selectivity with better flow characteristics. Cross-linked Sepharose gels are resistant to organic solvents and are thus the choice for separations in organic solvents.