rmp Protein A Sepharose™ Fast Flow Affinity Purification Media, GE Healthcare
Supplier: GE Healthcare - Life Sciences
Sepharose™
17513802 17513801 17513803 17513801 17513803 17513802
97067-898EA 776.34 USD InStock
CA97067-900 CA97067-898 CA97067-902 97067-898 97067-902 97067-900
rmp Protein A Sepharose™ Fast Flow Affinity Purification Media, GE Healthcare
Protein Purification Media
rmp Protein A Sepharose Fast Flow is a low leakage, non-mammalian based affinity medium designed for high purity separation of monoclonal and polyclonal antibodies at the laboratory and process scales.
  • Multi-point attachment of rProtein A by chemically-stable amide bonds
  • Coupling chemistry that minimizes leakage of protein A ligand
  • Reductive Amination coupling method
  • Stable in aqueous buffers commonly used in protein A chromatography – 10 mM HCl (pH 2), 1 mM NaOH (pH 11), 0.1 M sodium citrate/HCl (pH 3), 6 M GuHCl, 20% ethanol
rmp Protein A Sepharose™ Fast Flow is composed of recombinant Protein A coupled to the well established Sepharose™ 4 Fast Flow base matrix by reductive amination resulting in multi-point attachment. Reductive amination creates chemically stable amide bonds and resulting low ligand leakage.
rmp Protein A Sepharose™ 4 Fast Flow is manufactured without using animal-derived components.
As a member of the BioProcess resins range, rmp Sepharose™ 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.