Magnetic Beads, TiO2 Mag Sepharose, Cytiva
28-9440-10 28-9513-77 28-9513-77 28-9440-10
89129-072EA 57.6 USD
CA89129-072 89129-072 CA89129-106 89129-106
Magnetic Beads, TiO2 Mag Sepharose, Cytiva
TiO₂ Mag Sepharose™ magnetic beads are designed for enrichment of phosphopeptides from tryptic digested protein samples.
  • Magnetic beads for selective capture and enrichment of phosphopeptides
  • Easy to use - visible and dense magnetic beads
  • Simple protocol with elution conditions optimized for MS analysis
  • Parallel handling of samples - six samples can be processed in less than one hour
TiO₂ Mag Sepharose™ magnetic beads are based on titanium dioxide (TiO₂) chromatography and are designed to simplify capture and enrichment of phosphopeptides. TiO₂ has high affinity for phosphopeptides and is an efficient method for enrichment of phosphopeptides from complex samples.
TiO₂ Mag Sepharose™ improves ease-of-use in sample preparation of phosphopeptides. The magnetic bead format has excellent properties for small-scale experiments. The high density of the beads allows rapid capture by magnetic devices while the visibility of the beads ensures reliable collection of all bound target peptides.
When used together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel.