Streptavidin HP SpinTrap Buffer kit comprises pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation using Streptavidin HP SpinTrap columns
- High quality buffers
- All buffer components listed and defined
- Convenient format - stock solutions can be diluted directly into the provided bottles for storage
- Eliminates time-consuming buffer preparation, which allows fast, reproducible protein enrichment
- Sufficient for performing 16 reactions
The buffers provided in the kit are optimized for standard affinity-based enrichment/immunoprecipitation according to the protocol provided with Streptavidin HP SpinTrap columns
Ordering information: Kit includes 10X Binding/Washing Buffer [TBS]: 0.5 M Tris, 1.5 M NaCl, pH 7.5; 10X Elution Buffer: 1 M glycine-HCl, pH 2.9; Blocking Buffer: 2 mM biotin; 50 mM Tris, 0,15 M NaCl, pH 7.5.