Would you like to log in again?
Anti-GST Antibody, GE Healthcare
Supplier: GE Healthcare - Life Sciences
27-4577-01 27-4577-01
95017-606EA 496.06 USD InStock
95017-606 CA95017-606L
Anti-GST Antibody, GE Healthcare
Antibodies
For the highly sensitive and specific detection of recombinant schistosomal GST fusion proteins expressed using pGEX vectors.
  • Extensively cross-adsorbed to remove antibodies that bind to E. coli proteins and affinity purified using Glutathione Sepharose
  • Recommended for use in Western blots and dot blots to detect GST fusion proteins
  • Because of its polyclonal nature, it can recognize more than one epitope on GST, thereby improving its capacity for recognizing GST fusion proteins even if some binding sites are masked due to recombinant protein folding
Anti-GST Antibody is a polyclonal antibody purified from the sera of goats immunized with purified schistosomal GST, because of its polyclonal nature, it can recognize more than one epitope on GST. Anti-GST Antibody is a polyclonal antibody purified from the sera of goats immunized with purified schistosomal GST. Because of its polyclonal nature, it can recognize more than one epitope on GST, thereby improving its capacity for recognizing GST fusion proteins even if some binding sites are masked due to recombinant protein folding.
Ordering information: Supplied unconjugated for use with any enzyme-conjugated anti-goat antibody.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty