Would you like to log in again?
Nucleon HT Genomic DNA Extraction Kit, GE Healthcare
Supplier: GE Healthcare - Life Sciences
RPN8509 RPN8509
95026-700EA 308 USD InStock
CA95026-700L 95026-700
Nucleon HT Genomic DNA Extraction Kit, GE Healthcare
Nucleic Acid Purification Systems and Kits Genomic DNA Purification Systems and Kits
The illustra Nucleon HT kit facilitates the simple, phenol-free extraction of genomic DNA from paraffin-embedded sections and hard tissue requiring proteinase K digestion.
  • Each kit is designed to recover high-quality DNA from 50 preparations of up to 25 mg of hard tissue
  • Consistent chloroform extraction of high-quality, amplifiable DNA from formalin-fixed, paraffin-embedded (FFPE) tissue sections
  • User-friendly protocol enables efficient processing of large numbers of samples
As DNA purified from FFPE tissue slides is highly degraded, damaged, and available only in small quantities, little molecular analysis has been performed on such samples. illustra Nucleon HT protocols have been designed to prepare high-quality DNA from hard tissues and FFPE tissue sections that is suitable for genomic DNA amplification. Together, these tools enable molecular analysis of genome integrity and DNA copy number changes from FFPE tissue slides.
The Nucleon proprietary resin is added following cell lysis, deproteinization with sodium perchlorate, and a single chloroform extraction. Aspiration of the top layer of nucleic acids is simplified and volume of usable sample maximized by a barrier formed between the two layers. The recovered DNA is suitable for a variety of molecular biology applications, including whole genome amplification.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty