Rapid-hyb Buffers, Cytiva
Supplier: Cytiva
RPN1635 RPN1635 RPN1636 RPN1636
95038-160EA 645.24 USD
95038-160 CA95038-158L 95038-158 CA95038-160L
Rapid-hyb Buffers, Cytiva
Buffers Electrophoresis Buffers Hybridization Buffers
A major limitation of membrane hybridization is speed. At commonly used probe concentrations (2 to 10 ng/ml), hybridization normally takes 16 hours to detect single-copy gene sequences in mammalian DNA. Use of Rapid-hyb Buffer reduces this time to 2 hours using similar probe concentrations.
Store at ambient temperature. Under these conditions the product is stable for at least 3 months.