Would you like to log in again?
Amersham Hybond-NX, GE Healthcare
Supplier: GE Healthcare - Life Sciences
Hybond™
RPN303T RPN303T RPN1520T RPN1520T RPN203T RPN203T
95038-412EA 435.84 USD InStock
CA95038-412L 95038-412 95038-350 CA95038-350L CA95038-386L 95038-386
Amersham Hybond-NX, GE Healthcare
Transfer Membranes
Amersham Hybond-NX is a neutral nylon membrane with all the properties of Hybond-N
It can be used for multiple re-probings due to its high tensile strength. Hybond-NX gives cleaner backgrounds than Hybond-N when using hybridization buffer volumes below 70 µl/cm² of membrane. The neutral nylon membrane provides a binding capacity for nucleic acid of 600 µg/cm², developed specifically for use with low hybridization buffer volumes and high-throughput applications. Highly recommended for nucleic acid transfer applications using radioactive detection, particularly DNA dot/slot blots and applications requiring low volume hybridization.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty