Would you like to log in again?
Amersham Hybond-N, GE Healthcare
Supplier: GE Healthcare - Life Sciences
JACKSON SAFETY® Hybond™
RPN3050N RPN1210N RPN137N RPN82N RPN203N RPN1210N RPN87N RPN82N RPN1520N RPN303N RPN137N RPN203N RPN87N RPN2020N RPN132N RPN132N RPN2020N RPN1520N RPN303N RPN3050N
95038-310PK 177.17 USD InStock
CA95038-416L 95038-310 95038-328 95038-430 CA95038-382L CA95038-310L CA95038-438L CA95038-430L CA95038-346L CA95038-408L CA95038-328L 95038-382 95038-438 95038-370 95038-318 CA95038-318L CA95038-370L 95038-346 95038-408 95038-416
Amersham Hybond-N, GE Healthcare
Transfer Membranes
Amersham™ Hybond™-N is a neutral nylon membrane with a binding capacity for nucleic acids up to 600 µg/cm².
  • Recommended for nucleic acid blotting and microarray applications
Amersham™ Hybond™-N is a strong, supported membrane that is inherently hydrophilic and requires no pre-wetting. Nucleic acids can be quickly and easily crosslinked to nylon using UV light. When using low buffer volumes Hybond™-NX is recommended.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty