Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Immobiline DryStrip Cover Fluid, GE Healthcare

Supplier: GE Healthcare - Life Sciences
17-1335-01 17-1335-01
95056-338EA 101.05 USD InStock
CA95056-338L 95056-338
Immobiline DryStrip Cover Fluid, GE Healthcare
Electrophoresis Reagents Blotting Reagents
This purified fraction of mineral oil protects immobiline drystrip gels from drying and from the atmosphere during the first dimension of 2-D electrophoresis

  • Boiling point at 330°C
  • Insoluble in cold and hot water
  • Storage conditions from 18 to 25 °C

The fluid is suitable as a contact between cooling plate and gel backing film in horizontal electrophoresis.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty