SE 400 Vertical Unit, Cytiva
Supplier: Cytiva
SE400-15-1.5 80-6155-24
95044-794EA 1703.33 USD
CA95044-794L 95044-794
SE 400 Vertical Unit, Cytiva
Electrophoresis Systems Vertical Gel Electrophoresis Systems
Economical and secure vertical slab electrophoresis units with many of the conveniences of SE 600 series units.
  • Built-in casting stand seals sandwiches leak-free without tape.
Rugged injection-molded construction for easy assembly and durability in high-use labs.
Plates, clamps, spacers and combs are interchangeable with SE 600 series units.
SE 400 Vertical Units run one 16-cm gel or two gels with an accessory divider plate. Its low cost makes the SE 400 appropriate for labs requiring multiple units when temperature control is not crucial. The unit base is also a convenient casting stand for single gels. To cast multiple gels for one or more units, use the multiple gel casters.
Certifications: The product fulfills valid directives and standards when used within the conditions specified in the user manual. The product must also be used in the same state as it was delivered from Cytiva and connected only to other CE labeled Cytiva modules or other products as recommended. For regulatory details please see declaration of conformity.