PhastGel Buffer Strips, Cytiva
Supplier: Cytiva
17051601 17-0516-01 17-0517-01
95017-161PK 104.29 USD
95017-161 CA95017-161L CA95016-869L
PhastGel Buffer Strips, Cytiva
PhastGel Buffer Strips are made of GelBond polyester film
PhastGel DNA Buffer Strips are tested for DNA separations. When used with PhastGel DNA Silver Staining Kit, they improve the sensitivity of detection.
They are available in three types - PhastGel DNA, PhastGel SDS and PhastGel Native.
Ordering information: Supplied as ready-to-use agarose blocks, making buffer preparation unnecessary.