Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
PhastGel Homogeneous, GE Healthcare
Supplier: GE Healthcare - Life Sciences
Phastgel™
17-0624-01 17-0624-01
95040-402PK 282.79 USD InStock
95040-402 CA95040-402L
PhastGel Homogeneous, GE Healthcare
Electrophoresis Gels Precast Gels
Separate proteins and peptides on denaturing, native, and isoelectric focusing gels.
  • Select from homogeneous and gradient polyacrylamide gels
  • Separation zone: 20% T/2% C
  • Stacking zone: 7.5% T/2% C
Save time and improve reproducibility with precast gels, buffer strips, and staining kits.
Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits.
PhastGel Homogeneous media are available in three concentrations of polyacrylamide as indicated by their names: PhastGel Homogeneous 7.5, 12.5, and 20 and PhastGel High Density.
PhastGel High Density is a gel with 20% Polyacrylamide optimized for the separation of peptides and small proteins with relative molecular weights below about 20 000. To increase resolution of molecules with Mr below 10 000, ethylene glycol (30%) has been added.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty