Would you like to log in again?
HE 33 Mini Submarine Unit, GE Healthcare
Supplier: GE Healthcare - Life Sciences
80-6052-64 80-6052-64 80-6052-45 80-6052-45
95044-754EA 546.16 USD InStock
95044-754 CA95044-754L CA95044-752L 95044-752
HE 33 Mini Submarine Unit, GE Healthcare
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
  • Coolant-insulated buffer chamber runs 7 x 10 cm agarose gels in less than 20 minutes
  • Convenient, leak-proof casting trays mean fast gel casting without tape
  • UV-transparent gel tray offers convenient gel handling and viewing
  • Combs allow up to 32 samples per run
Unit has an enhanced passive cooling system, allowing you to run gels at high voltages for excellent speed and resolution in a mini-gel submarine unit. Convenient, leak-proof casting trays mean fast gel casting without tape. HE 33 Mini Submarine Unit has an enhanced passive cooling system, allowing you to run gels at high voltages for excellent speed and resolution in a mini-gel submarine unit. The novel molded base is designed to be filled with a common coolant such as 50% ethylene glycol so that it readily maintains short-term cooling capacity after chilling in a freezer. This allows for highly efficient heat exchange.
HE 33 Mini Submarine Unit has an enhanced passive cooling system, allowing you to run gels at high voltages for excellent speed and resolution in a mini-gel submarine unit. The novel molded base is designed to be filled with a common coolant such as 50% ethylene glycol so that it readily maintains short-term cooling capacity after chilling in a freezer. This allows for highly efficient heat exchange.
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty