Would you like to log in again?
Agarose NA, GE Healthcare
Supplier: GE Healthcare - Life Sciences
17-0554-03 17-0554-01 17-0554-01 17-0554-03 17-0554-02 17-0554-02
95040-296EA 2196 USD InStock
95040-296 CA95040-292L 95040-292 CA95040-296L CA95040-294L 95040-294
Agarose NA, GE Healthcare
Electrophoresis Gels Agarose and Acrylamide
Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications
Suitable for all analytical separations, as well as most preparative methods. If separated nucleic acids are to be used as substrates for enzymes (ligase and restriction endonucleases), a special highly purified agarose such as Agarose NA should be used.
For preparative work, an agarose with low gelling temperature should be used. Agarose Prep has low melting and gelling points.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty