Select-a-Dye Mono-Reactive Dye Packs, Cytiva
PA15123 PA15123
95056-512EA 514.55 USD
95056-512 CA95056-512L
Select-a-Dye Mono-Reactive Dye Packs, Cytiva
Enables a variety of labeling strategies using NHS ester, maleimide, or hydrazide to be investigated.
  • Available as either Cy3 or Cy5
  • Example protocols included with each dye pack
  • Contains 0.5 mg of each labeling chemistry per pack