Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amersham Hypercassette Autoradiography Cassettes, GE Healthcare

Supplier: GE Healthcare - Life Sciences
RPN11644 RPN11645 RPN11642 RPN11643 RPN11650 RPN11648 RPN11649 RPN11647
Amersham hypercssette autoradiography cassettes are designed to survive the toughest demands of research in the life sciences

  • Robust and chip-proof autoradiography cassettes
  • Light-tight exposures at temperatures as low as -70°C
  • Maximum contact between film and sample provides optimal resolution
  • Nonporous pad gives a wipe-clean surface ensuring minimum contamination

Amersham Hypercassette autoradiography cassettes are constructed from strong aluminium and designed for the toughest demands of life science research. Deep cassettes are 1.5 cm in depth, allowing them to accommodate thicker samples. A choice of Hypercassette colors enables color coding of work. Use a permanent marker to record details on the polycarbonate label and be confident of secure identification, then wipe clean after use.




 

ORDER

Expand All / Collapse All
Description Gel Size Supplier No. VWR Catalog Number Unit Price Quantity
Hypercassette, Neutral (Standard) 8 x 10" RPN11649 95039-986 Each Retrieving
Hypercassette, Neutral (Standard) 5 x 7" RPN11648 95039-984 Each Retrieving
Product available on GSA Advantage®
Hypercassette, Neutral (Standard) 18 x 24 cm RPN11642 95039-972 Each Retrieving
Product available on GSA Advantage®
Hypercassette, Neutral (Standard) 10 x 12" RPN11650 95039-988 Each Retrieving
Product available on GSA Advantage®
Hypercassette, Neutral (Standard) 24 x 30 cm RPN11643 95039-974 Each Retrieving
Product available on GSA Advantage®
Hypercassette, Neutral (Standard) 20 x 40 cm RPN11647 95039-982 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Depth 0.75 cm

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty