Would you like to log in again?
Amersham Amplify Fluorographic Reagent, GE Healthcare
Supplier: GE Healthcare - Life Sciences
NAMP100 NAMP100
95040-024EA 470.31 USD InStock
CA95040-024L 95040-024
Amersham Amplify Fluorographic Reagent, GE Healthcare
Electrophoresis Reagents Molecular Weight Markers
Amplify™ is a fast and convenient fluorographic reagent for ³⁵S-, ¹⁴C- and ³H-labelled samples in gels. Amplify™ can increase sensitivity of detection and significantly reduce exposure times required for weak beta emitters.
  • Increase detection efficiency 1000-fold for ³H and 15-fold for ³⁵S and ¹⁴C
  • Less than 2 hours of total preparation time
  • Can be used with acrylamide or agarose gels with no change in protocols
  • Odourless and inorganic
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty