Would you like to log in again?
PIG® Spill Kit in Cart, New Pig
KIT217
56620-108EA 578.81 USD InStock
56620-108
PIG® Spill Kit in Cart, New Pig
Spill Control Materials
Excellent for maintenance crews, as well as labs or other areas prone to small spills. The 8" rubber wheels roll easily over uneven surfaces. PIG Blue Socks stop spreading spills; PIG Mat and Pillows absorb; wipers clean surfaces and hands; PIG Repair Putty can patch a drum leak or fix a crack in a fluid reservoir.
  • Excellent for maintenance crews, labs or other areas prone to small spills
  • 8" rubber wheels roll easily over uneven surfaces
  • Socks stop spreading spills; Mat and Pillows absorb; Wipers clean surfaces and hands; Repair Putty patch leaks or cracks
  • Temporary disposal bags, gloves, goggles, and boot covers make clean-up easier
Temporary disposal bags help make cleanup easier
Optional Refill for PIG® Spill Kit in Cart is packed to refill your PIG Spill Kit. PIG Blue Socks stop spreading spills; PIG Mat and Pillows absorb; wipers clean surfaces and hands; PIG Repair Putty can patch a drum leak or fix a crack in a fluid reservoir. Temporary disposal bags help make cleanup easier. Includes gloves, goggles and boot covers to guard against splashes.
Includes gloves, goggles and boot covers to guard against splashes. Prepacked, compartmentalized cart provides easy access to contents. Narrow width fits easily through doors and aisles. How-to instructions are printed on the cabinet doors. Lockable for added security. Top and bottom handles for easy carrying. Not UN rated.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty