Peptide Vessel with Ace-Thred Top and Bottom, Ace Glass Incorporated
Supplier: Ace Glass
6408-06 6408-08 6408-04 6408-10 6408-02
89054-240EA 133.56 USD
89054-240 89054-242 89054-244 89054-246 89054-248
Peptide Vessel with Ace-Thred Top and Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Cylindrical vessels with #15 Ace-Thred at both ends
Vessels have no side port. Use PTFE adapter (89052-602 - not included) with removable filter disc and PTFE valve (89052-608 - not included) with Luer-Lok extension at either end.