Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
VWR® General-Purpose Freestanding Undercounter Laboratory Freezer
VWR®
SCUCFS-0420
89425-622EA 824 USD InStock
89425-622
VWR® General-Purpose Freestanding Undercounter Laboratory Freezer
Freezers Laboratory / General Purpose Freezers
The VWR® General-Purpose Freestanding Undercounter Laboratory Freezer is the ideal choice for laboratories and pharmacies requiring undercounter or countertop applications with adequate space for ventilation.
  • VWR® two-year parts and labor warranty
  • VWR® five-year compressor parts warranty
Freezers are supplied preset at –20 °C (–4 °F)
The unit requires a minimum of 10.2 cm (4") of air flow space in the back, and 7.6 cm (3") on both sides of the unit for proper heat removal. Failure to provide the proper space for air circulation will void any and all warranty claims.
These economical units are intended for general-purpose applications only, where precise temperature control and maintenance are not essential and there are no requirements for temperature alarms.
The unit also includes leveling legs, keyed door locks, and an 0.95 cm (3/8") probe access port. This freezer features a solid door and white exterior finish.
Ordering information: Accessory items must be purchased separately.
Delivery information: Freezers are supplied with a two-year parts and labor warranty, with an additional three-year warranty on compressor parts.
Order Now
ORDER
ACCESSORIES
COMPLEMENTARY PRODUCTS
SERVICES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty