Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PIG® Oil-Only Absorbent Mat Pad in Dispenser Box, New Pig

Supplier: NEW PIG CORP
MAT485
Eight layers of 100% polypropylene are thermally bonded to give PIG Mat excellent strength; won't rip, tear or fray even when saturated

Can be wrung out and incinerated after use to reduce waste or for fuels blending. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Dispenser box makes mat easy to access, carry and store while protecting contents from dirt and moisture. Pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need, so you use less mat and save money.

Lightweight construction is economical for small tasks and cleanups.

Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue. Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water. Bright white color makes absorbed oil easier to see; draws attention to machine leaks and clearly shows saturation level during spill response. Floats to clean up spills on water.




 

ORDER

Expand All / Collapse All
Description Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
Lightweight, 200 Pads 38.1 x 50.8 cm (15 x 20") MAT485 89094-960 Case of 200 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty