Sarcophine ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

BVT-0305-M002 BVT-0305-M010
102513-740EA 131.54 USD
102513-740 102513-742
Sarcophine ≥97% (by HPLC)
Sarcophine

Acetylcholine esterase and ATPase inhibitor. Ichthyotoxin. Tumorigenesis and JB6 cell transformation inhibitor.

Formula: C₂₀H₂₈O₃
Storage Temperature: Freezer
CAS Number: 55038-27-2

Order Now

Specification Test Results

Source/Host Isolated from Sarcophyton glaucum.
Purity 97% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in methanol or dichloromethane.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR