Aureothin ≥96% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

BVT-0303-M001 BVT-0303-M005
102513-736EA 304.62 USD
102513-736 102513-738
Aureothin ≥96% (by HPLC)
Aureothin

Oxidoreductase inhibitor. Antitrypanosomal, antibacterial, antifungal, insecticidal and pesticidal. Antitumor compound.

Formula: C₂₂H₂₃NO₆
Storage Temperature: Freezer
CAS Number: 2825-00-5
UN: 3462
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces thioluteus.
Purity 96% (HPLC)
Appearance/Color Yellow solid.
Solubility Soluble in 100% ethanol, methanol, DMSO, dimethylformamide, dicholormethane or acetone.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR