Decarestrictine D ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

BVT-0283-M001
102513-708EA 124.62 USD
102513-708
Decarestrictine D ≥98% (by HPLC)
Decarestrictine D

Hypolipidemic. Cholesterol biosynthesis inhibitor.

Formula: C₁₀H₁₆O₅
Storage Temperature: Freezer
CAS Number: 127393-89-9

Order Now

Specification Test Results

Source/Host Isolated from Penicillium sp.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, methanol or acetone.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR