Curvulin ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

BVT-0097-M001
102513-540EA 180 USD
102513-540
Curvulin ≥98% (by HPLC)
Curvulin

Phytotoxin. Microtubule assembly inhibitor. Similar to curvularin.

Formula: C₁₂H₁₄O₅
MW: 238,2
Storage Temperature: Freezer
CAS Number: 19054-27-4

Order Now

Specification Test Results

Source/Host Isolated from Curvularia sp.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in methanol, DMSO or acetone.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR