Hypothemycin ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

BVT-0067-M001 BVT-0067-C250
102513-498EA 152.31 USD
102513-498 102513-500
Hypothemycin ≥98% (by HPLC)
Hypothemycin

Antifungal. Cytotoxic against some tumor cell lines, partly attributed to inhibition of Ras-inducible genes. Inhibits proliferation of mouse and human T cells. Modulates production of cytokines during T cell activation. Facilitates the ubiquitinylation process of cyclin D1. Potent and selective threonine/tyrosine-specific kinase, MEK and other protein kinases inhibitor in both in vitro and in vivo studies.

Formula: C₁₉H₂₂O₈
Storage Temperature: Freezer
CAS Number: 76958-67-3

Order Now

Specification Test Results

Source/Host Isolated from Phoma sp.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or acetone; insoluble in methanol or water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR