Resistomycin ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

BVT-0036-M001
102513-484EA 131.54 USD
102513-484
Resistomycin ≥98% (by HPLC)
Resistomycin

Antibacterial (Gram-positive and mycobacteria). RNA polymerase inhibitor. Apoptosis inducer.

Formula: C₂₂H₁₆O₆
MW: 376,4
Storage Temperature: Freezer
CAS Number: 20004-62-0

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces sp.
Purity 98% (HPLC)
Appearance/Color Yellow to orange solid.
Solubility Soluble in DMSO or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR