Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decoyinine ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

Warning

BVT-0030-M001
102513-474EA 96.92 USD InStock
102513-474
Decoyinine ≥98% (by NMR)
Decoyinine
Nucleoside antibiotic. Antitumor compound. Xanthosine monophosphate (XMP) aminase inhibitor. RNA synthesis inhibitor. Specific GMP synthase inhibitor. Reduces intracellular GTP levels.


Formula: C₁₁H₁₃N₅O₄
MW: 279,3
Storage Temperature: Freezer
CAS Number: 2004-04-8
Order Now
Specification Test Results
Source/Host Isolated from Streptomyces sp. S 2113.
Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO (warm), water or 100% ethanol: poorly soluble in methanol.


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty