Demecolcine ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
Colcemid
Anti-cancer compound. Cell synchronization agent. Used as a drug in chemotherapy by synchronizing tumor cells at metaphase, the radiosensitive stage of the cell cycle. Microtubule assembly inhibitor. Depolymerizes microtubules and limits microtubule formation (inactivates spindle fibre formation). Inhibits mitosis during cell division at metaphase by inhibiting spindle formation. Low concentrations affect microtubule dynamics and cell migration, while high concentrations promote microtubule detachment from microtubule organizing center. Apoptosis inducer. Thrombopoietic agent.
Formula:
C₂₁H₂₅NO₅ MW: 371.433 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00075459 CAS Number: 477-30-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Source/Host | Semisynthetic from colchicine (Prod. No. AG-CN2-0048). |
Purity |
|
Appearance/Color | White to off-white powder. |
Solubility | Soluble in ethanol, DMSO or chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...