Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

Avarol ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS


AGCN20044M001
102515-828EA 173.08 USD InStock
102515-828
Avarol ≥97% (by HPLC)
Avarol
Antibacterial against selected Gram-positive strains, antifungal. Anticancer compound. Antileukemic. Antimitotic agent. Antiviral compound. Inhibits HIV activity. Anti-inflammatory. Synovial phospholipase A2, cyclooxygenase and lipoxygenase inhibitor. Modulator of superoxide dismutase and glutathione peroxidase. Platelet aggregation inhibitor. Anti-psoriatic, mediated by inhibition of TNF-alpha generation and NF-kappaB activation. Reviews.


Formula: C₂₁H₃₀O₂
MW: 314,5
Storage Temperature: Freezer
CAS Number: 55303-98-5
Order Now
Specification Test Results
Source/Host Isolated from sponge Dysidea avara.
Purity 97% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in ethanol or DMSO.


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty