Jasplakinolide ≥98% (by HPLC), high purity

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0037-C100 AG-CN2-0037-C050
102515-810EA 540 USD
102515-810 102515-808
Jasplakinolide ≥98% (by HPLC), high purity
Jasplakinolide

Cell permeable, non-fluorescent F-actin probe. Potent inducer of actin polymerization and stabilization. Competes with phallotoxins for actin binding. Antifungal and antiparasitic compound. Antiproliferative and anticancer compound. Apoptosis enhancer/inducer. Tool used for autophagy/phagocytosis research.

Formula: C₃₆H₄₅BrN₄O₆
Storage Temperature: Freezer
CAS Number: 102396-24-7

Order Now

Specification Test Results

Source/Host Isolated from Jaspis splendens.
Purity 98% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in ethanol, methanol or DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR