PP1 ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
1-(tert-Butyl)-3-(p-tolyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
AG-CR1-3562-M005
AG-CR1-3562-M001
102516-280EA
62.31
USD
102516-280
102516-282
PP1 ≥98% (by HPLC)
PP1
Highly potent and selective Scr family tyrosine kinase inhibitor. Shows anti-Ras cancer potential by blocking Ras-induced activation of PAK1. Antitumor compound. RIP2 inhibitor. Blocks TGF-beta-mediated cellular responses.
Formula:
C₁₆H₁₉N₅ MW: 281.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01076570 CAS Number: 172889-26-8 |
Specification Test Results
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO. Slightly soluble in ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...