Paprotrain ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-3504-M001
102516-224EA 124.62 USD
102516-224
Paprotrain ≥98% (by NMR)
Paprotrain

Cell permeable and reversible kinesin-specific MKLP-2 (mitotic kinesin-like protein 2) inhibitor. Inhibits the recruitment of the chromosome passenger proteins survivin and aurora B to the central spindle during anaphase chromosome segregation. Moderate DYRK1A inhibitor (IC50= 5.5 µM). Lead compound.

Formula: C₁₆H₁₁N₃
MW: 245.3
Storage Temperature: Freezer
MDL Number: MFCD00617529
CAS Number: 57046-73-8

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Yellow solid.
Solubility Soluble in DMSO (at least 5 mg/ml).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR