Bisindolylmaleimide IX·methanesulfonate ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
Synonyms:
Ro 31-8220 mesylate
AG-CR1-0111-M005
AG-CR1-0111-M001
102516-160EA
228.46
USD
102516-160
102516-158
Bisindolylmaleimide IX·methanesulfonate ≥98% (by NMR)
Bisindolylmaleimide IX·methanesulfonate
Selective and cell permeable protein kinase C (PKC) inhibitor. Inhibits the stimulation of insulin secretion by glucose. Inhibits T cell activation. Apoptosis inducer. Potent glycogen synthase kinase-3 (GSK-3) inhibitor. Transcription inhibitor. Induces TNF receptor family-mediated cell death. Pim-1 kinase inhibitor. Antitumor compound.
Formula:
C₂₅H₂₃N₅O₂S·CH₄O₃S MW: 457.6·96.1 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03788207 CAS Number: 138489-18-6 |
Specification Test Results
Purity |
|
Appearance/Color | Red solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...