Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sirtinol ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS


AGCR10055M005 AGCR10055M001 AGCR10055M025
102516-090EA 76.15 USD InStock
102516-090 102516-088 102516-092
Sirtinol ≥98% (by NMR)
Sirtinol
Specific cell permeable SIRT1 (sirtuin 1) inhibitor. Senescence-like growth arrest inducer. Platelet aggregation inhibitor. Apoptosis inducer.


Formula: C₂₆H₂₂N₂O₂
MW: 394,46
Storage Temperature: Refrigerator
MDL Number: MFCD00810186
CAS Number: 410536-97-9
Order Now
Specification Test Results
Purity 98% (NMR)
Appearance/Color Off-white to yellow solid.
Solubility Soluble in DMSO, 100% ethanol or dimethyl formamide.


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty