Dynasore ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CR1-0045-M005
AG-CR1-0045-M025
102516-058EA
221.54
USD
102516-058
102516-056
Dynasore ≥98% (by NMR)
Dynasore hydrate
Dynamin-1, -2 (DNM1, DNM2) and Dynamin-related protein 1 (DRP1) inhibitor. Endocytosis inhibitor. Viral infection inhibitor. Destabilizes actin filaments. Inhibits entry of Trypanosoma into macrophages. Inhibitor of mitophagy. Inhibitor of hepatocyte lipophagy
Formula:
C₁₈H₁₄N₂O₄·0.5H₂O Storage Temperature: Freezer |
CAS Number:
1202867-00-2 UN: 3077 ADR: 9,III |
Specification Test Results
Purity |
|
Appearance/Color | Off-white solid. |
Solubility | Soluble in 100% ethanol (1 mg/ml) or DMSO (10 mg/ml); insoluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...