AMI-1 ≥99% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

AG-CR1-0038-M005 AG-CR1-0038-M025
102516-030EA 443.08 USD
102516-030 102516-028
AMI-1 ≥99% (by NMR)
AMI-1

Potent, cell permeable and specific protein arginine N-methyltransferase 1 (PRMT1) inhibitor (IC50 = 8.8 µM). Does not compete for the AdoMet (S-adenosyl-L-methionine; SAM) binding site. Blocks peptide substrate binding. HIV-1 RT polymerase inhibitor (IC50 = 5.0 µM).

Formula: C₂₁H₁₂N₂O₉S₂Na₄
MW: 592.4 g/mol
Storage Temperature: Freezer
CAS Number: 20324-87-2

Order Now

Specification Test Results

Purity 99% (NMR)
Appearance/Color Brown solid.
Solubility Soluble in water, 100% ethanol or DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR