PK 11195 ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CR1-0008-M050
AG-CR1-0008-M010
102515-896EA
276.92
USD
102515-896
102515-894
PK 11195 ≥98% (by HPLC)
PK 11195
Selective peripheral benzodiazepine antagonist. Apoptosis enhancer. Glucose-induced insulin secretion inhibitor. Induces mitochondria cytochrome c release. Anticancer compound. Antiproliferative. Pharmacological tool in autophagy.
Formula:
C₂₁H₂₁ClN₂O Storage Temperature: Refrigerator |
CAS Number:
85532-75-8 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in 100% ethanol, methanol, DMSO or dichloromethane; insoluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...