Pellitorine ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: (2E,4E)-N-Isobutyldeca-2,4-dienamide

AG-CN2-0009-M005 AG-CN2-0009-M001
102515-690EA 124.62 USD
102515-690 102515-688
Pellitorine ≥97% (by HPLC)
Pellitorine

Tingling-inducing agent. Excellent stable model compound for sensory studies. Exerts same profile as the unstable compound hydroxy-alpha-sanshool. Shows larvicidal, antimycobacterial and antituberculosis activity. alpha-Glucosidase inhibitor used in diabetes mellitus, cancer, infection and inflammatory research. ACAT (Acyl-CoA cholesteryl acyl transferase) inhibitor. Potential anti-cancer lead compound. Anti-thrombotic. Anti-septic. Antiprotozoal, antimalarial activity.

Formula: C₁₄H₂₅NO
MW: 223.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01735995
CAS Number: 18836-52-7

Order Now

Specification Test Results

Source/Host Synthetic. Originally isolated from roots of Anacyclus pryrethrum and fruits of Piper nigrum.
Purity 97% (HPLC)
Appearance/Color Yellow to beige solid.
Solubility Soluble in 100% ethanol, methanol, DMSO or dichloromethane. Sparingly soluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR