Anti-NLRP12/NALP12 Rabbit Polyclonal Antibody

AG25B0021C100
102514-682EA 401.54 USD
102514-682
Anti-NLRP12/NALP12 Rabbit Polyclonal Antibody
NALP12/NLRP12 mediates activation of caspase-1 via Asc and promotes activation of NF-kappa-B via IKK. It is involved in the building of an inflammasome complex. Defects in NALP12/NLRP12 are the cause of familial cold autoinflammatory syndrome type 2 (FCAS2).

Stable for at least 1 year after receipt when stored at -20°C.

Type:
Antigen: NLRP12/NALP12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR