Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PIG® Drive-Over DrainBlocker® Drain Covers, New Pig

Supplier: NEW PIG CORP
PLR300 PLR301 PLR302 PLR303 PLR304 PLR305 PLR306 PLR307 PLR308 PLR309
Excellent for long-term drain protection where liquid transfers, washdowns or other leak- and spill-prone jobs are common

Tough, patented-rubber HogHyde Skin stands up to punctures, tearing and abrasion, even under heavy vehicle traffic from tankers, trucks or forklifts. Super-sealing urethane bottom layer conforms to uneven surfaces and tightly seals drain openings to block liquid.

Withstands UV rays, infrared light and all weather conditions; keep outdoors as long as needed without worrying about degradation. Polyurethane and rubber materials resist water, oils and many chemicals. Leave down as preventive measure or store for emergency response.

Eliminates the need to frequently deploy and remove a temporary cover or permanently seal your drains.




 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
For Rectangular Drains up to 48 x 24" PLR309 89107-552 Each Retrieving
For Rectangular Drains up to 54 x 12" PLR307 89107-548 Each Retrieving
For Rectangular Drains up to 54 x 30" PLR308 89107-550 Each Retrieving
For Square Drains up to 12" PLR300 89107-534 Each Retrieving
For Square Drains up to 18" PLR301 89107-536 Each Retrieving
For Square Drains up to 24" PLR302 89107-538 Each Retrieving
For Square Drains up to 30" PLR303 89107-540 Each Retrieving
For Square Drains up to 36" PLR304 89107-542 Each Retrieving
For Square Drains up to 42" PLR305 89107-544 Each Retrieving
For Square Drains up to 48" PLR306 89107-546 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping