Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Rose Bengal Agar with Chloramphenicol, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
H35 W87 H35 W87
89407-858PK 67.91 USD InStock
CA89407-858 CA89407-862 89407-858 89407-862
Rose Bengal Agar with Chloramphenicol, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and enumeration of yeasts and molds
Many yeasts and molds will not grow in an acidic pH environment. This media has a neutral pH which helps grow yeast and mold, but is also conducive to the growth of bacteria. The addition of chloramphenicol helps reduce bacterial overgrowth.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty