Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Mineral Oil, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z80 Z79 Z79
76213-938EA 104.68 USD InStock
76213-938 CA89407-734 89407-734
Mineral Oil, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For oil overlays.
Used as a sealant to create an anaerobic environment for biochemical tests such as decarboxylase; and oxidation and fermentation reactions.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty