Would you like to log in again?
Corn Meal Agar with Tween® 80, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G101 G101
89407-134PK 63.38 USD InStock
CA89407-134 89407-134
Corn Meal Agar with Tween® 80, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of fungi
Induces chlamydospore formation by Candida spp.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty