CRITERION™ Nutrient Gelatin is used to aid in classification of the Enterobacteriaceae by determining an organism's ability to liquefy gelatin. Dehydratedculture media with engineered container.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
· Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
· Convenient handgrip - 'no slip' finger holds
· Stackable containers - for efficient storage
· Increase solubility - dissolves quickly
· Custom and bulk products available to your specifications
· Tested for optimal performance
· BSE Free (contact for specifics)
· Over 30 years of formulation experience
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
Custom and bulk products available
BSE Free documentation available. Over 30 years of formulation experience
Certifications: BSE Free