Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Hektoen Enteric Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™
C5842 C5843 C5842 C5843 C5841 C5840 C5841 C5840
89405-484EA 1394.81 USD InStock
CA89405-484 CA89405-486 89405-484 89405-486 CA89405-482 89405-480 89405-482 CA89405-480
Hektoen Enteric Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Hektoen Enteric (HE) Agar is a selective and differential medium used for the isolation and differentiation of gram-negative enteric pathogens
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty